KEY003448527 Key Organics / BIONET

ethyl 6-chloroimidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD02186220
  • Cas Number : 67625-38-1
  • Molecular Weight : 224.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112 - 114
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72766

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0683_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart