KEY003448528 Key Organics / BIONET

ethyl 3-bromoimidazo[1,2-a]pyrimidine-2-carboxylate

  • MDL number MFCD02186429
  • Cas Number : 134044-63-6
  • Molecular Weight : 270.086

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201 - 203
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72767

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0684_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart