KEY003448905 Key Organics / BIONET

ethyl 2,4-dimethyl-7-phenylpyrrolo[1,2-a]pyrimidine-8-carboxylate

  • MDL number MFCD00171951
  • Cas Number : 338419-86-6
  • Molecular Weight : 294.354

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135 - 138
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74379

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4F-964_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart