KEY003449109 Key Organics / BIONET

6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD02186559
  • Cas Number : 182181-19-7
  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228 - 229
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76539

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0623_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart