KEY003449121 Key Organics / BIONET

methyl 4-(1,1-dioxo-1lambda6-thiomorpholin-4-yl)benzoate

  • MDL number MFCD02661674
  • Cas Number : 45185-76-0
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154 - 156
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76568

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0669_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart