KEY003449235 Key Organics / BIONET

1-(1,3-thiazol-2-yl)-1H-pyrrole-2-carbonitrile

  • MDL number MFCD03012931
  • Cas Number : 439108-81-3
  • Molecular Weight : 175.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58 - 59
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76890

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0600_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart