KEY003449271 Key Organics / BIONET

4-methyl-5-{2-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazol-4-yl}-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03012595
  • Cas Number : 955962-78-4
  • Molecular Weight : 346.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76951

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0828_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart