KEY003449388 Key Organics / BIONET

5-bromo-2-oxo-1-{[3-(trifluoromethyl)phenyl]methyl}-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD01935940
  • Cas Number : 338982-36-8
  • Molecular Weight : 376.129

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187 - 188
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78096

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5F-393S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart