KEY003449611 Key Organics / BIONET

3-[3-(2,5-dimethyl-1H-pyrrol-1-yl)thiophen-2-yl]-1H-pyrazol-5-amine

  • MDL number MFCD02661683
  • Cas Number : 1030622-14-0
  • Molecular Weight : 258.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184 - 186
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80124

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-0053_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart