KEY003449833 Key Organics / BIONET

2-(cyanomethyl)-5,7-bis(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD00141799
  • Cas Number : 338786-45-1
  • Molecular Weight : 319.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138 - 140
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81581

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6F-051_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart