KEY003449894 Key Organics / BIONET

7-(thiophen-2-yl)-[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

  • MDL number MFCD00232507
  • Cas Number : 338793-17-2
  • Molecular Weight : 217.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252 - 254
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81940

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6H-035_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart