KEY003450112 Key Organics / BIONET

2,4-dibromo-6-methyl-3-(propan-2-yl)phenol

  • MDL number MFCD02571954
  • Cas Number : 70454-10-3
  • Molecular Weight : 308.013

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83729

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0600_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart