KEY003450113 Key Organics / BIONET

1-(6-chloro-2-methylpyridin-3-yl)ethan-1-one

  • MDL number MFCD02661714
  • Cas Number : 439111-18-9
  • Molecular Weight : 169.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83745

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0618_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart