KEY003450519 Key Organics / BIONET

1-{6-methyl-[1,2,4]triazolo[3,2-b][1,3]thiazol-5-yl}ethan-1-one

  • MDL number MFCD01814533
  • Cas Number : 187597-18-8
  • Molecular Weight : 181.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147 - 149
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86432

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7M-582S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart