KEY003450673 Key Organics / BIONET

3-(4-methylbenzamido)thiophene-2-carboxylic acid

  • MDL number MFCD02572044
  • Cas Number : 478080-03-4
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197 - 200
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87620

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7R-1333_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart