KEY003450685 Key Organics / BIONET

3-amino-4-methylpentanoic acid

  • MDL number MFCD00800505
  • Cas Number : 5699-54-7
  • Molecular Weight : 131.175

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204 - 206
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87658

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7T-0026_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart