KEY003450705 Key Organics / BIONET

3-methyl-1,3-thiazolidine-2,4-dione

  • MDL number MFCD02194457
  • Cas Number : 16312-21-3
  • Molecular Weight : 131.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40 - 43
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87696

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7T-0094_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart