KEY003450712 Key Organics / BIONET

1,4,6-trimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00829059
  • Cas Number : 64038-03-5
  • Molecular Weight : 162.192

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202 - 204
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87708

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7T-0120_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart