KEY003450760 Key Organics / BIONET

7-cyclopropylpyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD03305760
  • Cas Number : 866132-04-9
  • Molecular Weight : 184.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165 - 167
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87773

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7T-0298_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart