KEY003450832 Key Organics / BIONET

6-{[3-(trifluoromethyl)phenyl]sulfanyl}imidazo[2,1-b][1,3]thiazole-5-carbaldehyde

  • MDL number MFCD00102587
  • Cas Number : 175277-56-2
  • Molecular Weight : 328.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86 - 89
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88493

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8D-077_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart