KEY003451153 Key Organics / BIONET

(2E)-3-(4-tert-butylphenyl)prop-2-enoic acid

  • MDL number MFCD00040719
  • Cas Number : 1208-65-7
  • Molecular Weight : 204.269

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195 - 197
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90690

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/8R-0658_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart