KEY003451260 Key Organics / BIONET

3-amino-4-methylhexanoic acid

  • MDL number MFCD03305808
  • Cas Number : 40469-87-2
  • Molecular Weight : 145.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190 - 192
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90877

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8T-0052_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart