KEY003451383 Key Organics / BIONET

4-chloro-[1,2,4]triazolo[4,3-a]quinoxaline

  • MDL number MFCD00140798
  • Cas Number : 62603-54-7
  • Molecular Weight : 204.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 288 - 291
Purity 95
Storage temp. Store under N2 at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91756

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9E-902_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart