KEY004181235 Key Organics / BIONET

4-(propan-2-yl)-1,2,3-thiadiazole-5-carboxylic acid

  • MDL number MFCD01935987
  • Cas Number : 183302-68-3
  • Molecular Weight : 172.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141 - 143
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1261

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10J-338S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart