KEY004181241 Key Organics / BIONET

2-(1,1-dioxo-1lambda6-thiomorpholin-4-yl)benzoic acid

  • MDL number MFCD03001215
  • Cas Number : 400878-25-3
  • Molecular Weight : 255.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227 - 229
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2508

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10P-213_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart