KEY004181243 Key Organics / BIONET

(2E)-3-[5-chloro-2-(prop-2-yn-1-yloxy)phenyl]prop-2-enoic acid

  • MDL number MFCD03001216
  • Cas Number : 695186-82-4
  • Molecular Weight : 236.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155 - 158
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2768

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0226_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart