KEY004181244 Key Organics / BIONET

[5-chloro-2-(prop-2-yn-1-yloxy)phenyl]methanol

  • MDL number MFCD03001217
  • Cas Number : 400751-45-3
  • Molecular Weight : 196.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55 - 58
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2775

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0233_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart