KEY004181252 Key Organics / BIONET

[3-methoxy-4-(prop-2-yn-1-yloxy)phenyl]methanol

  • MDL number MFCD03001222
  • Cas Number : 385383-48-2
  • Molecular Weight : 192.214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58 - 60
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2943

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0812_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart