KEY004181254 Key Organics / BIONET

3-[2-(dimethylamino)ethyl]-4-imino-6,7-dimethoxy-1,2,3,4-tetrahydroquinazoline-2-thione

  • MDL number MFCD03001223
  • Cas Number : 400751-59-9
  • Molecular Weight : 308.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2945

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0815_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart