KEY004181255 Key Organics / BIONET

4-methoxy-3-(prop-2-yn-1-yloxy)benzaldehyde

  • MDL number MFCD03001224
  • Cas Number : 145654-01-9
  • Molecular Weight : 190.198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76 - 77
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2962

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0844_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart