KEY004181257 Key Organics / BIONET

3-amino-4-[(2,6-dichlorophenyl)sulfanyl]benzonitrile

  • MDL number MFCD01935934
  • Cas Number : 338982-10-8
  • Molecular Weight : 295.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 180 - 184
Iupac Name 3-amino-4-[(2,6-dichlorophenyl)sulfanyl]benzonitrile
Inchi Code 1S/C13H8Cl2N2S/c14-9-2-1-3-10(15)13(9)18-12-5-4-8(7-16)6-11(12)17/h1-6H,17H2
Inchi Key FISQHGPGQXANAC-UHFFFAOYSA-N

Description

Melting Point 180 - 184
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5243

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/11K-550S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart