KEY004181265 Key Organics / BIONET

1-[3-(4-chlorophenyl)-1,2-oxazol-5-yl]ethan-1-ol

  • MDL number MFCD03425717
  • Cas Number : 109572-24-9
  • Molecular Weight : 223.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63 - 67
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5438

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11L-518S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart