KEY004181266 Key Organics / BIONET

1-(prop-2-en-1-yl)-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00224218
  • Cas Number : 830-74-0
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 87 - 90
Iupac Name 1-allyl-1H-indole-2,3-dione
Inchi Code 1S/C11H9NO2/c1-2-7-12-9-6-4-3-5-8(9)10(13)11(12)14/h2-6H,1,7H2
Inchi Key ZWNYDPBLEDGGQD-UHFFFAOYSA-N

Description

Melting Point 87 - 90
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5474

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/11L-570S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart