KEY004181267 Key Organics / BIONET

2-(phenylmethanimidamido)acetic acid

  • MDL number MFCD01935985
  • Cas Number : 32683-07-1
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 210
Iupac Name {[imino(phenyl)methyl]amino}acetic acid
Inchi Code 1S/C9H10N2O2/c10-9(11-6-8(12)13)7-4-2-1-3-5-7/h1-5H,6H2,(H2,10,11)(H,12,13)
Inchi Key ZDTNJTKQGYFSCI-UHFFFAOYSA-N

Description

Melting Point 210
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5712

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/11M-317S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart