KEY004181275 Key Organics / BIONET

3-(2,6-dimethylmorpholin-4-yl)-1,1,1-trifluoropropan-2-ol

  • MDL number MFCD03001226
  • Cas Number : 667938-72-9
  • Molecular Weight : 227.227

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6532

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11P-647_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart