KEY004181278 Key Organics / BIONET

methyl 3-methoxy-2,8-dioxo-1-oxaspiro[4.5]deca-3,6,9-triene-4-carboxylate

  • MDL number MFCD03001190
  • Cas Number : 316354-15-1
  • Molecular Weight : 250.206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120 - 122
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6630

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11R-0062_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart