KEY004181282 Key Organics / BIONET

ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD01833013
  • Cas Number : 353258-31-8
  • Molecular Weight : 292.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132 - 134
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6679

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11R-0255_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart