KEY004181285 Key Organics / BIONET

methyl 2-{[(2,4-dichlorophenyl)methyl]sulfanyl}benzoate

  • MDL number MFCD03001231
  • Cas Number : 308851-38-9
  • Molecular Weight : 327.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114 - 117
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6690

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11R-0273_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart