KEY004181286 Key Organics / BIONET

2-{[(2,4-dichlorophenyl)methyl]sulfanyl}benzoic acid

  • MDL number MFCD03001232
  • Cas Number : 385383-42-6
  • Molecular Weight : 313.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217 - 219
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6698

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11R-0286_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart