KEY004181287 Key Organics / BIONET

methyl 2-{[(3-fluorophenyl)methyl]sulfanyl}benzoate

  • MDL number MFCD03001233
  • Cas Number : 385383-54-0
  • Molecular Weight : 276.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70 - 72
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6699

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11R-0287_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart