KEY004181290 Key Organics / BIONET

2-(3-chloropropyl)-3-(4-methoxyphenyl)-1-oxo-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acid

  • MDL number MFCD03001235
  • Cas Number : 385383-38-0
  • Molecular Weight : 373.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192 - 195
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6710

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-0306_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart