KEY004181292 Key Organics / BIONET

2-cyclopentyl-3-(2-methoxyphenyl)-1-oxo-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acid

  • MDL number MFCD03001237
  • Cas Number : 385383-40-4
  • Molecular Weight : 365.429

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198 - 200
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6717

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11R-0325_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart