KEY004181293 Key Organics / BIONET

3-(4-chlorophenyl)-2-cyclohexyl-1-oxo-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acid

  • MDL number MFCD03001191
  • Cas Number : 400073-91-8
  • Molecular Weight : 383.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197 - 199
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6719

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11R-0329_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart