KEY004181299 Key Organics / BIONET

methyl 4-acetyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD02571735
  • Cas Number : 40611-82-3
  • Molecular Weight : 167.164

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118 - 120
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6782

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11R-0827_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart