KEY004181305 Key Organics / BIONET

2-(4-chlorobenzenesulfonamido)propanoic acid

  • MDL number MFCD00706949
  • Cas Number : 90410-27-8
  • Molecular Weight : 263.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145 - 147
Purity 95
Storage temp. RT
Physical Form White to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8087

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12G-396S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart