KEY004181307 Key Organics / BIONET

2-(4-chlorobenzenesulfonamido)acetic acid

  • MDL number MFCD01104937
  • Cas Number : 13029-72-6
  • Molecular Weight : 249.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 180 - 182
Iupac Name {[(4-chlorophenyl)sulfonyl]amino}acetic acid
Inchi Code 1S/C8H8ClNO4S/c9-6-1-3-7(4-2-6)15(13,14)10-5-8(11)12/h1-4,10H,5H2,(H,11,12)
Inchi Key LOZHABFZMAUWBU-UHFFFAOYSA-N

Description

Melting Point 180 - 182
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8100

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12G-418S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart