KEY004181313 Key Organics / BIONET

methyl 5-chloro-4-formyl-1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD01109226
  • Cas Number : 401571-11-7
  • Molecular Weight : 264.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123 - 126
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8916

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12M-534S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart