KEY004181314 Key Organics / BIONET

methyl 5-[(4-chlorophenyl)sulfanyl]-4-formyl-1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD05021848
  • Cas Number : 449778-53-4
  • Molecular Weight : 372.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109 - 112
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8921

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12M-541S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart