KEY004181315 Key Organics / BIONET

3-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzaldehyde

  • MDL number MFCD03425720
  • Cas Number : 439094-98-1
  • Molecular Weight : 253.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 40 - 42
Iupac Name 3-[(2-chloro-1,3-thiazol-5-yl)methoxy]benzaldehyde
Inchi Code 1S/C11H8ClNO2S/c12-11-13-5-10(16-11)7-15-9-3-1-2-8(4-9)6-14/h1-6H,7H2
Inchi Key ZPJNAWYNZXASEW-UHFFFAOYSA-N

Description

Melting Point 40 - 42
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8981

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-008_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart