KEY004181318 Key Organics / BIONET

4-[(2,4-dichlorophenyl)methoxy]benzaldehyde

  • MDL number MFCD00610864
  • Cas Number : 70627-17-7
  • Molecular Weight : 281.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102 - 104
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9340

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-044_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart